95680 Fluka

p-Xylene

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: 1,4-Dimethylbenzene

  • CAS Number 106-42-3

  • Linear Formula C6H4(CH3)2

  • Molecular Weight 106.17

  •  Beilstein Registry Number 1901563

  •  EC Number 203-396-5

  •  MDL number MFCD00008556

  •  PubChem Substance ID 24890185

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Aromatics,
grade   analytical standard
vapor density   3.7 (vs air)
vapor pressure   9 mmHg ( 20 °C)
assay   ≥99.5% (GC)
autoignition temp.   984 °F
shelf life   (limited shelf life, expiry date on the label)
expl. lim.   7 %
refractive index   n20/D 1.495(lit.)
  n20/D 1.495
bp   138 °C(lit.)
mp   12-13 °C(lit.)
density   0.861 g/mL at 20 °C(lit.)

Description

Packaging

5, 50 mL in glass bottle

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

<I>m</I>-Xylene

analytical standard

<I>o</I>-Xylene

analytical standard

<I>p</I>-Xylene

puriss. p.a., ≥99.0% (GC)

<I>p</I>-Xylene
296333p-Xylene

anhydrous, ≥99%

<I>p</I>-Xylene
317195p-Xylene

CHROMASOLV®, for HPLC, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?