Synonym: Xylene mixture of isomers
CAS Number 1330-20-7
Linear Formula C6H4(CH3)2
Molecular Weight 106.17
MDL number MFCD00077264
Popular Documents: FTNMR (PDF)
| vapor pressure | 18 mmHg ( 37.7 °C) |
| grade | purum |
| assay | ≥97.0% (total assay of C8H10-isomers, GC) |
| expl. lim. | 7 % |
| impurities | ~20% ethylbenzene |
| refractive index |
n |
|
n | |
| bp | 137-140 °C(lit.) |
Sales restrictions may apply
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?