Synonym: Pentafluorophenyl oxalate
CAS Number 16536-48-4
Empirical Formula (Hill Notation) C14F10O4
Molecular Weight 422.13
Beilstein Registry Number 2068395
MDL number MFCD00143774
PubChem Substance ID 24890274
| product line | BioReagent |
| suitability | suitable for chemiluminescence |
Chemiluminescence: Emission: 430 nm (reaction of TCPO with 9,10-Diphenylanthracene and H2O2)
Reagent for a sensitive detn. method of fluorescent compds. by peroxyoxalate ester chemiluminescence in HPLC and CE1,2
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?