Synonym:
3,4′,5,7-Tetrahydroxyflavone,
3,5,7-
CAS Number 520-18-3
Empirical Formula (Hill Notation) C15H10O6
Molecular Weight 286.24
Beilstein Registry Number 304401
EC Number 208-287-6
MDL number MFCD00016938
| Related Categories | Aesculus, Allium, Alphabetical, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard |
| assay | ≥99.0% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| storage temp. | 2-8°C |
Chromogenic reagent for antimony in the low ppm range2 and for gallium and indium in the sub-ppm range.3
Potent inhibitor of osteoclastic bone resorption. The effect is believed to be attributable to both the antioxidant and estrogenic activities of kaempferol.1
Customers Also Viewed
≥95% (HPLC), solid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?