Synonym: Stannic sulfate in sulfuric acid
Linear Formula Sn(SO4)2 · H2SO4
MDL number MFCD01863137
PubChem Substance ID 24890318
| Related Categories | Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Puriss p.a., T-Z, Puriss p.a. More... |
| grade | puriss. p.a. |
| form | suspension |
| quality | for the determination of nitrate acc. to DIN 38405-D9-3 |
| impurities | 0.22-0.26% Sn |
| ~80% H2SO4 | |
| density | 1.74 g/mL at 20 °C |
precipitation has to be agitated
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?