97580 Fluka

Mueller-Hinton Agar 2

for microbiology

DOWNLOAD MSDS (PDF)

Synonym: M-H 2 Agar

Properties

Related Categories A - M, All Solid Media/Agars, Bacteria (General Media), Clinical Media, Enterococcus,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
composition   agar, 17.0 g/L
  beef heart infusion, 2 g/L
  casein acid hydrolysate, 17.5 g/L
  starch, soluble, 1.5 g/L
pH   7.3±0.2 (25 °C)
suitability   Haemophilus species
  nonselective for Bacteria (General Media)
  nonselective for Enterococcus
  nonselective for Escherichia coli and Coliforms
  nonselective for Pseudomonas
  nonselective for Staphylococcus
  nonselective for Streptococcus

Description

Application

Mueller Hinton Agar 2 is a solid medium recommended by Bauer et al. and NCCLS for susceptibility testing of pathogens. Antimicrobial susceptibility test discs impregnated with known amount of antibiotics are placed on the agar surface. Manufactured to contain low levels of thymine, thymidine, calcium and magnesium.

Price and Availability

Customers Also Viewed

for microbiology

for microbiology

for microbiology, Cation-adjusted

for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?