B2926 Fluka

Brucella Agar with Hemin and Vitamin K1

for microbiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories A - M, All Solid Media/Agars, Anaerobic Bacteria, Bacteroides, Brucella,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
pH   7.0±0.2 (25 °C)
suitability   Anaerobic Bacteria
  nonselective for Bacteroides
  nonselective for Brucella
  nonselective for Clostridia

Description

Components

Components (g/L):
Casein Enzymic Hydrolysate 10.00
Peptic Digest of Animal Tissue 10.00
Yeast Extract 2.00
Dextrose 1.00
Sodium Chloride 5.00
Sodium Bisulphite 0.10
Hemin 0.10
Vitamin K1 0.01
Agar 15.00

Application

Used for the isolation and subculture of anaerobes.

Price and Availability

Customers Also Viewed

for microbiology

for microbiology

Hemin
51280Hemin

BioXtra, from Porcine, ≥98.0% (HPLC)

Hemin
H9039Hemin

from bovine, ≥90%

Vitamin K<SUB>1</SUB>

viscous liquid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?