CAS Number 8024-48-4
MDL number MFCD00212912
Popular Documents: Product Information Sheet (PDF)
| Related Categories | Aloe Vera, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, C, |
| storage temp. | 2-8°C |
Total hydroxyanthracene derivatives ≥20%, calculated as cascaroside A; ≥80% of the total hydroxyanthracene derivatives are cascarosides, calculated as cascaroside A.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?