C9095 Fluka

Casein yeast magnesium agar

for microbiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories A - M, All Solid Media/Agars, Escherichia coli & Coliforms, Media, Media by Microorganism,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
final pH   7.0±0.2 (25 °C)
suitability   nonselective for Escherichia coli and Coliforms

Description

Components

Components in g/L:
Casein enzymic hydrolysate, 10.00
Yeast extract, 5.00
Sodium chloride, 5.00
Magnesium sulfate, 0.98
Agar, 15.00

Application

Recommended for use in the cultivation of recombinant strains of E. coli.

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?