D2560 Fluka

DNase test agar with toluidine blue

for microbiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories A - M, All Food Control Media (alphabetical), All Solid Media/Agars, Bacteria (General Media), Biochemical Identification Media by Test,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
final pH   7.3±0.2 (25 °C)
suitability   nonselective and differential for Bacteria (General Media)
  nonselective and differential for Molds and Yeasts (General Media)
  nonselective and differential for Serratia
  nonselective and differential for Staphylococcus
  nonselective and differential for Streptococcus

Description

Components

Ingredients (g/L)
Tryptose, 20.00
Deoxyribonucleic acid (DNA), 2.00
Sodium chloride, 5.00
Toluidine blue, 0.10
Agar, 15.00

Application

Recommended for the detection of deoxyribonuclease activity of bacteria and fungi with the indicator toluidine blue.

Price and Availability

Customers Also Viewed

for microbiology

for microbiology

Dimethyl sulfoxide

Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%

Toluidine Blue

for microscopy (Hist., Vit.)

Toluidine Blue O

Technical grade

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?