L5910 Fluka

Lysine medium

for microbiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories A - M, All Liquid Media/Broths, Biochemical Identification Media by Test, Biochemical Identification Media, A to Z, Candida,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
final pH   5.0±0.2 (25 °C)
suitability   nonselective for Candida
  nonselective for Pichia
  nonselective for Saccharomyces
  nonselective for Zygosaccharomyces

Description

Components

Ingredients (g/L)
Dextrose, 44.50
Monopotassium phosphate, 1.78
Magnesium sulphate, 0.89
Calcium chloride, 0.178
Sodium chloride, 0.089
Adenine, 0.00178
DL-Methionine, 0.000891
L-Histidine, 0.000891
DL-Tryptophane, 0.000891
Boric acid, 0.0000089
Zinc sulphate, 0.0000356
Ammonium molybdate, 0.0000178
Manganese sulphate, 0.0000356
Ferrous sulphate, 0.0002225
L-Lysine, 1.00
Inositol, 0.02
Calcium pantothenate, 0.002
Aneurine, 0.0004
Pyridoxine, 0.0004
p-Aminobenzoic acid, 0.0002
Nicotinic acid, 0.0004
Riboflavin, 0.0002
Biotin, 0.000002
Folic acid, 0.000001
Agar, 17.80

Application

Used for isolation and enumeration of wild yeasts in pitching yeasts.

Price and Availability

Customers Also Viewed

for microbiology

Potassium <SC>L</SC>-lactate solution

~60%, for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?