Synonym:
5-
CAS Number 31431-39-7
Empirical Formula (Hill Notation) C16H13N3O3
Molecular Weight 295.29
Beilstein Registry Number 759809
EC Number 250-635-4
MDL number MFCD00057872
PubChem Substance ID 24896742
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Antibiotics, Antibiotics A to Z, |
| assay | ≥98% (NT/TLC) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?