Synonym: Corn starch, Starch from maize
CAS Number 9005-25-8
Linear Formula (C6H10O5)n
EC Number 232-679-6
MDL number MFCD00082026
PubChem Substance ID 24899646
Popular Documents: Bulletin (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Reagents, Analytical Standards, Analytical/Chromatography, Application Specific Reagents, |
| storage temp. | 2-8°C |
Customers Also Viewed
sufficient for 20 assays
sufficient for 20 assays


Unmodified waxy corn starch of essentially pure amylopectin; contains only trace amounts of amylose.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?