Synonym:
N1-
CAS Number 80-35-3
Empirical Formula (Hill Notation) C11H12N4O3S
Molecular Weight 280.30
Beilstein Registry Number 277076
MDL number MFCD00057372
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, S, SN - SZ More... |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?