Synonym:
4-Amino-N-
CAS Number 723-46-6
Empirical Formula (Hill Notation) C10H11N3O3S
Molecular Weight 253.28
Beilstein Registry Number 6732984
EC Number 211-963-3
MDL number MFCD00010546
PubChem Substance ID 24899752
Popular Documents: Product Information Sheet (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, S, SN - SZ More... |
| Gene Information | rat ... Ednra(24326) |
Sulfonamide antibiotic that blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase.
Mode of Action: Inhibits folic acid synthesis in prokaryotes.
Anti-microbial Spectrum: Gram positive, Gram negative, Chlamydia
Mode of Resistance: Alteration of dihydropteroate synthase or alternative pathway for folic acid synthesis.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?