Synonym:
1-
CAS Number 64-77-7
Empirical Formula (Hill Notation) C12H18N2O3S
Molecular Weight 270.35
Beilstein Registry Number 1984428
EC Number 200-594-3
MDL number MFCD00027169
PubChem Substance ID 24277742
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Application Index, Bioactive Small Molecules, |
| Gene Information |
human ... ALB(213), CYP1A2(1544), CYP2C9(1559), EPHX2(2053), KCNJ1(3758) mouse ... Ephx2(13850) |
Anti-diabetic agent. Metabolized by CYP2C9 (tolbutamide hydroxylase).
Customers Also Viewed
≥97%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?