T5509 Fluka

Trigonelline hydrochloride

DOWNLOAD MSDS (PDF)

Synonym: 1-Methylpyridinium-3-carboxylate hydrochloride, N-Methylnicotinic acid betaine hydrochloride

  • CAS Number 6138-41-6

  • Empirical Formula (Hill Notation) C7H7NO2 · HCl

  • Molecular Weight 173.60

  •  Beilstein Registry Number 3734169

  •  EC Number 228-119-5

  •  MDL number MFCD00077250

  •  PubChem Substance ID 24900287

Price and Availability

Customers Also Viewed

Betaine
B2629Betaine

≥98% (perchloric acid titration)

Chlorogenic acid

≥95% (titration)

Nicotinic acid

≥98%

Nicotinic acid

≥99.5% (HPLC)

Trigonelline hydrochloride

purum, ≥98.0% (AT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?