Synonym:
N-
CAS Number 33978-72-2
Empirical Formula (Hill Notation) C21H29NO4 · HCl
Molecular Weight 395.92
MDL number MFCD00153884
PubChem Substance ID 24277815
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Structure, Chromatography, |
| grade | analytical standard, for drug analysis |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?