B3426 Fluka

Bile salts mixture

for microbiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories Base Ingredients for Media, Bile, Microbiology More...
grade   for microbiology

Description

Application

Used as an inhibitory agent of gram positive bacteria in microbiological culture media.

Preparation Note

Extracted from purified, fresh bile.

Price and Availability

Customers Also Viewed

dried, unfractionated

for microbiology

for microbiology

for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?