Synonym: Aflatoxin M1 solution
CAS Number 6795-23-9
Beilstein Registry Number 1630643
MDL number MFCD00871812
PubChem Substance ID 24892227
| Related Categories | A, AA to AL, All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, |
| grade | BCR® certified Reference Material |
| concentration | 9.93 μg/mL in chloroform |
| storage temp. | 2-8°C |
material produced by BCR®
Sealed ampule containing ~2.5 ml
BCR is a registered trademark of European Commission
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?