Synonym: Flumequine and oxolinic acid in lyophilised salmon tissue
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, S, SA - SM More... |
| grade | BCR® certified Reference Material |
| storage temp. | −70°C |
BCR is a registered trademark of European Commission
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?