Synonym: Peptone from casein
CAS Number 91079-40-2
EC Number 293-428-4
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Base Ingredients for Media, Broth / Media, Core Bioreagents, Life Science Reagents for Cloning, Media Components, |
| biological source | from bovine milk |
| nitrogen analysis | 6.0-7.0% amino, ≥11.0% total |
Recommended for the production of media for the growth of Charon phage and other lambda phages.
Recommended concentration: 25 g/L
N-Z-Amine is a registered trademark of Kerry Group
Customers Also Viewed
for microbiology
Casein enzymatic hydrolysate, from bovine milk
plant cell culture tested
Casein enzymatic hydrolysate, from bovine milk
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?