CAS Number 521-35-7
Empirical Formula (Hill Notation) C21H26O2
Molecular Weight 310.43
EC Number 200-659-6
MDL number MFCD00056774
PubChem Substance ID 24892874
Popular Documents: Datasheet (PDF)
| Related Categories | β-Glucuronidase, β-Glucuronidase Drug Analysis Bundle, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical and Industrial Enzymes, |
| grade | analytical standard, for drug analysis |
| assay | ≥98% |
| drug control | Home Office Schedule 1 |
| concentration | 1.0 mg/mL in methanol |
| 1.0 mg/mL±5% in methanol | |
| storage temp. | 2-8°C |
| Gene Information |
human ... CNR2(1269) rat ... Cnr1(25248) |
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?