Synonym: Peptone from casein
CAS Number 91079-40-2
EC Number 293-428-4
| Related Categories | Base Ingredients for Media, Microbiology, Protein Sources (Peptones & Extracts), Protein Sources (derived from animal tissue), Protein Sources A to Z More... |
| grade | for microbiology |
| form | powder |
| nitrogen analysis | ≥4.6% amino |
| ≥7% total | |
| ash | ≤42% |
| loss | ≤5% loss on drying |
| color | off-white to yellow |
Casein Acid hydrolysate vitamin free is specially standardized for the purpose of pertussis vaccine production, but may be used for any vaccines/toxins production where standardized vitamin concentrations are needed.
A hydrochloric acid digest of casein free of vitamins.
Customers Also Viewed
Casein acid hydrolysate, from bovine milk
technical grade
powder
for microbiology
for microbiology
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?