C8845 Fluka

Casein hydrolysate broth

for microbiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories A - M, All Liquid Media/Broths, Enterococcus, Media, Media by Application,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
final pH   7.3±0.2 (25 °C)
suitability   nonselective for Enterococcus
  nonselective for Staphylococcus

Description

Components

Components g/L:
Casein acid hydrolysate, 20.00
Ferric citrate, 0.025
Potassium phosphate, 2.00
Magnesium sulfate, 0.20
L-Cystine, 0.025
Sodium acetate, 7.00
L-Tryptophan, 0.075
Calcium pantothenate, 0.0005
Thiamine, 0.00004
Nicotinic acid, 0.0012

Application

Used for production of stapholococcal enterotoxin.

Price and Availability

Customers Also Viewed

for microbiology

Casein enzymatic hydrolysate, from bovine milk

from bovine milk, Casein enzymatic hydrolysate

powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?