• USA Home
  • CL22 - CCS Viscosity Standard

EMAIL THIS PAGE TO A FRIEND

CL22 Fluka

CCS Viscosity Standard

ASTM D 5293 and SAE Specification J314

Synonym: Cold-Cranking Simulator Viscosity Standard

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, CCS Viscosity Standards, Paragon Scientific, Physical Properties Standards,
grade   certified reference material
shelf life   limited shelf life, expiry date on the label
mfr. no.   Paragon Scientific Ltd.
format   mixture

Description

General description

Find detailed technical data for all Paragon CCS Viscosity Standards on the Paragon Scientific webpage

This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO Guide 34 under UKAS accreditation

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Xn
Risk Statements (Europe) 
65
Safety Statements (Europe) 
62
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?