Empirical Formula (Hill Notation) C15H24O5
Molecular Weight 284.35
MDL number MFCD00274495
Popular Documents: Specification Sheet (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, D, DID - DIN, |
| assay | ≥97% (TLC) |
| form | solid |
| storage temp. | 2-8°C |
Antimalarial; antipsoriasis; anti-inflammatory.
artemisia plant extract
Protect from light.
Customers Also Viewed
from Artemisia annua
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?