EMAIL THIS PAGE TO A FRIEND

D7770 Fluka

Diminazene aceturate

analytical standard

Synonym: Berenil

Purchase

Description

Packaging

1 g in poly bottle

Price and Availability

Customers Also Viewed

Imidocarb dipropionate

VETRANAL, analytical standard

Nafamostat mesylate

≥98% (HPLC), solid

Netropsin dihydrochloride

from Streptomyces netropsis, ≥98% (HPLC and TLC), powder

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
MB7605000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Diminazene attenuates pulmonary hypertension and improves angiogenic progenitor cell functions in experimental models. Shenoy V, Gjymishka A, Jarajapu YP, et al. Am. J. Respir. Crit. Care Med. 187(6), 648-57, (2013)

Loading...


Cerebroprotection by angiotensin-(1-7) in endothelin-1-induced ischaemic stroke. Mecca AP, Regenhardt RW, O'Connor TE, et al. Exp. Physiol. 96(10), 1084-96, (2011)

Loading...


Detection of multiple drug-resistant Trypanosoma congolense populations in village cattle of south-east Mali. Mungube EO, Vitouley HS, Allegye-Cudjoe E, et al. Parasit. Vectors 5, 155, (2012)

Loading...


Inhibition of spermidine/spermine N1-acetyltransferase activity: a new therapeutic concept in rheumatoid arthritis. Neidhart M, Karouzakis E, Jüngel A, et al. Arthritis Rheumatology 66(7), 1723-33, (2014)

Loading...


A validated and stability indicating HPLC method for analysis of diminazene aceturate and antipyrine combination in a ready injectable solution. Abualhasan MN, Batrawi N, Zaid AN, et al. Drug Res. (Stuttg.) 63(6), 300-4, (2013)

Loading...


Angiotensin-converting enzyme 2 activation for treatment of pulmonary hypertension. Veit F and Weissmann N Am. J. Respir. Crit. Care Med. 187(6), 569-71, (2013)

Loading...


Biochemical and morphological changes in Trypanosoma brucei brucei-infected rats treated with homidium chloride and diminazene aceturate. Adeyemi OS and Sulaiman FA J. Basic Clin. Physiol. Pharmacol. 23(4), 179-83, (2012)

Loading...


Energetics of ligand binding to the DNA minor groove. Kostjukov VV, Hernandez Santiago AA, Rojas Rodriguez F, et al. Phys. Chem. Chem. Phys. 14(16), 5588-600, (2012)

Loading...


The effects of combination of methanolic leaf extract of Azadirachta indica and diminazene diaceturate in the treatment of experimental Trypanosoma brucei brucei infection in rats. Omoja VU, Anaga AO, Obidike IR, et al. Asian Pac. J. Trop. Med. 4(5), 337-41, (2011)

Loading...


Field detection of resistance to isometamidium chloride and diminazene aceturate in Trypanosoma vivax from the region of the Boucle du Mouhoun in Burkina Faso. Sow A, Sidibé I, Bengaly Z, et al. Vet. Parasitol. 187(1-2), 105-11, (2012)

Loading...


Hematologic and bone marrow changes in dogs experimentally infected with Rangelia vitalii. França RT, Da Silva AS, Costa MM, et al. Vet. Clin. Pathol. 42(1), 31-9, (2013)

Loading...


Antiglaucomatous effects of the activation of intrinsic Angiotensin-converting enzyme 2. Foureaux G, Nogueira JC, Nogueira BS, et al. Invest. Ophthalmol. Vis. Sci. 54(6), 4296-306, (2013)

Loading...


Serial haematology results in transfused and non-transfused dogs naturally infected with Babesia rossi. Scheepers E, Leisewitz AL, Thompson PN, et al. J. S. Afr. Vet. Assoc. 82(3), 136-43, (2011)

Loading...


Genital lesions in male red fronted gazelles (Gazella rufifrons) experimentally infected with Trypanosoma brucei and the effect of melarsamine hydrochloride (Cymelarsan®) and diminazene aceturate (Berenil®) in its treatment. Mbaya AW, Nwosu CO, and Kumshe HA Theriogenology 76(4), 721-8, (2011)

Loading...


Highly debilitating natural Trypanosoma vivax infections in Brazilian calves: epidemiology, pathology, and probable transplacental transmission. Batista JS, Rodrigues CM, Olinda RG, et al. Parasitol. Res. 110(1), 73-80, (2012)

Loading...


Purification of plasmid DNA from clarified and non-clarified Escherichia coli lysates by berenil pseudo-affinity chromatography. Caramelo-Nunes C, Gabriel MF, Almeida P, et al. J. Chromatogr. B. Analyt. Technol. Biomed. Life Sci. 904, 81-7, (2012)

Loading...


Involvement of mitochondrial genes of Babesia gibsoni in resistance to diminazene aceturate. Wickramasekara Rajapakshage BK, Yamasaki M, Hwang SJ, et al. J. Vet. Med. Sci. 74(9), 1139-48, (2012)

Loading...


CBFB and MYH11 in inv(16)(p13q22) of acute myeloid leukemia displaying close spatial proximity in interphase nuclei of human hematopoietic stem cells. Weckerle AB, Santra M, Ng MC, et al. Genes Chromosomes Cancer 50(9), 746-55, (2011)

Loading...


Diminazene diaceturate for treatment of chronic Cytauxzoon felis parasitemia in naturally infected cats. Lewis KM, Cohn LA, Marr HS, et al. J. Vet. Intern. Med. 26(6), 1490-3, (2012)

Loading...


Evaluation of in vitro and in vivo inhibitory effects of fusidic acid on Babesia and Theileria parasites. Salama AA, Aboulaila M, Moussa AA, et al. Vet. Parasitol. 191(1-2), 1-10, (2013)

Loading...


Pharmacokinetics of diminazene diaceturate in healthy cats. Lewis KM, Cohn LA, Birkenheuer AJ, et al. J. Vet. Pharmacol. Ther. 35(6), 608-10, (2012)

Loading...


Is trypanocidal drug resistance a threat for livestock health and production in endemic areas? Food for thoughts from Sahelian goats infected by Trypanosoma vivax in Bobo Dioulasso (Burkina Faso). Vitouley HS, Sidibe I, Bengaly Z, et al. Vet. Parasitol. 190(3-4), 349-54, (2012)

Loading...


Trypanocidal activity of the essential oils in their conventional and nanoemulsion forms: in vitro tests. Baldissera MD, Da Silva AS, Oliveira CB, et al. Exp. Parasitol. 134(3), 356-61, (2013)

Loading...


Analysis of energy generation and glycolysis pathway in diminazene aceturate-resistant Babesia gibsoni isolate in vitro. Rajapakshage BK, Yamasaki M, Murakami M, et al. Jpn. J. Vet. Res. 60(2-3), 51-61, (2012)

Loading...


High prevalence of drug resistance in animal trypanosomes without a history of drug exposure. Chitanga S, Marcotty T, Namangala B, et al. PLoS Negl. Trop. Dis. 5(12), e1454, (2011)

Loading...


Diminazene aceturate (Berenil) modulates the host cellular and inflammatory responses to Trypanosoma congolense infection. Kuriakose S, Muleme HM, Onyilagha C, et al. PLoS ONE 7(11), e48696, (2012)

Loading...


Validation of an HPLC method for the simultaneous determination of diminazene diaceturate and phenazone in injectable veterinary granules and bulk powders. Kassaye L, Hymete A, Bekhit AA, et al. Pak. J. Pharm. Sci. 25(1), 255-9, (2012)

Loading...


Merck 14,3266

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?