EMAIL THIS PAGE TO A FRIEND

E2499900 Fluka

Etofenamate

European Pharmacopoeia (EP) Reference Standard

Synonym: 2-[[3-(Trifluoromethyl)phenyl]amino]benzoic acid 2-(2-hydroxyethoxy)ethyl ester

  • CAS Number 30544-47-9

  • Empirical Formula (Hill Notation) C18H18F3NO4

  • Molecular Weight 369.34

Purchase

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Other Notes

Sales restrictions may apply.

Packaging

Unit quantity: 40 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference subtance catalogue.

Price and Availability

Customers Also Viewed

Bemotrizinol

analytical standard

European Pharmacopoeia (EP) Reference Standard

European Pharmacopoeia (EP) Reference Standard

Flufenamic acid

analytical standard, for drug analysis

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Allergic contact dermatitis from etofenamate: report of 9 cases. Hergueta JP, Ortiz FJ, and Iglesias L Contact Dermatitis 31(1), 60-2, (1994)

Loading...


Turpentine sensitization in a nonsteroidal anti-inflammatory solution user. Borrego L, Hernandez N, Martel R, et al. Dermatitis 23(4), 182-3, (2012)

Loading...


Action spectrum for etofenamate photoallergic contact dermatitis. Kerr A, Becher G, Ibbotson S, et al. Contact Dermatitis 65(2), 117-8, (2011)

Loading...


[Allergic contact eczema to etofenamate and dwarf pine oil]. Knöll R, Ulrich R, and Spallek W Sportverletz. Sportschaden 4(2), 96-8, (1990)

Loading...


[Intramuscular etofenamate in the treatment of acute lumbago. Effectiveness and tolerance in comparison with intramuscular diclofenac-Na]. Stratz T Fortschr. Med. 108(13), 264-6, (1990)

Loading...


Study on the effect of etofenamate 10% cream in comparison with an oral NSAID in strains and sprains due to sports injuries. Vanderstraeten G and Schuermans P Acta Belg. Med. Phys. 13(3), 139-41, (1990)

Loading...


Etofenamate associated with Lyell syndrome: a case report. Golcuk Y, Oray D, Atilla OD, et al. Clin. Toxicol. (Phila.) 48(5), 471-2, (2010)

Loading...


Allergic and photoallergic contact dermartitis from etofenamate: study of 14 cases. Goday Buján JJ, Pérez Varela L, Piñeyro Molina F, et al. Contact Dermatitis 61(2), 118-20, (2009)

Loading...


Allergic contact dermatitis to etofenamate. Cross-reaction to other nonsteroidal anti-inflammatory drugs. Alcántara Villar M, Pagan JA, Palacios L, et al. Contact Dermatitis 58(2), 118-9, (2008)

Loading...


[Etofenamate and the analgesic effect in the management of acute pain from spine in the emergency room]. Patiñio JL, Mejía SN, and Ramos EQ Acta Ortop. Mex. 21(5), 253-5, (2007)

Loading...


[Comparative clinical multicenter study to evaluate analgesic effectiveness of intramuscular etofenamate and diclofenac in patients with post-surgical pain]. Guevara-López U, Uscanga-Sánchez S, Márquez J, et al. Cir. Cir. 72(6), 483-90, (2004)

Loading...


Contact dermatitis from etofenamate. Götze A, Teikemeier G, and Goerz G Contact Dermatitis 26(3), 209, (1992)

Loading...


Contact dermatitis due to etofenamate. Guerra L, Piraccini BM, Adami F, et al. Contact Dermatitis 26(3), 199, (1992)

Loading...


[Plasma- and tissue concentrations following intramuscular administration of etofenamat. Pharmacokinetics of etofenamat and flufenamic acid in plasma, synovium, and tissues of patients with chronic polyarthritis after administration of an oily solution of etofenamat]. Köhler G, Tressel W, Dell HD, et al. Arzneimittelforschung 42(12), 1487-91, (1992)

Loading...


Combined contact and photocontact allergic dermatitis to etofenamate in flogoprofen gel. Sánchez-Pérez J, Sánchez TS, and García-Díez A Am. J. Contact Dermatitis 12(4), 215-6, (2001)

Loading...


Assessing the removal of pharmaceuticals and personal care products in a full-scale activated sludge plant. Salgado R, Marques R, Noronha JP, et al. Environ. Sci. Pollut. Res. Int. 19(5), 1818-27, (2012)

Loading...


Effects of some pharmacological agents on the survival of unipedicled venous flaps: an experimental study. Askar I, Saray A, Gurlek A, et al. Microsurgery 21(8), 350-6, (2001)

Loading...


Photoallergic contact dermatitis due to flufenamic acid and etofenamate. Montoro J, Rodriguez-Serna M, Liñana JJ, et al. Contact Dermatitis 37(3), 139-40, (1997)

Loading...


Allergic contact dermatitis from etofenamate without cross-sensitization to other anthranilic acid derivatives. Chu CY, Chen YL, Lin LJ, et al. Dermatology 206(4), 341-2, (2003)

Loading...


Etofenamate levels in human serum and synovial fluid following iontophoresis. Bender T, Bariska J, Rojkovich B, et al. Arzneimittelforschung 51(6), 489-92, (2001)

Loading...


[Allergic contact dermatitis caused by etofenamate]. Degenhardt A, Zick C, and Hausen BM Z. Hautkr. 63(6), 475-8, (1988)

Loading...


Influence of preemptive analgesia on pulmonary function and complications for laparoscopic cholecystectomy. Şen M, Özol D, and Bozer M Dig. Dis. Sci. 54(12), 2742-7, (2009)

Loading...


Comparative study of etofenamate and fentanyl for outpatient extracorporeal shockwave lithotripsy. Unsal A, Cimentepe E, Bozoklu A, et al. Scand. J. Urol. Nephrol. 35(6), 502-4, (2001)

Loading...


The isolated perfused bovine udder as an in vitro model of percutaneous drug absorption. Skin viability and percutaneous absorption of dexamethasone, benzoyl peroxide, and etofenamate. Kietzmann M, Löscher W, Arens D, et al. J. Pharmacol. Toxicol. Methods 30(2), 75-84, (1993)

Loading...


Topical etofenamate to facilitate venipuncture. Gulcu N, Karaaslan K, and Ugur F Pediatr. Emerg. Care 23(10), 759, (2007)

Loading...


Adverse reaction of topical etofenamate: petechial eruption. Orbak Z, Yildirim ZK, Sepetci O, et al. West Indian Med. J. 61(7), 767-9, (2012)

Loading...


Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?