ERMBF416C Fluka

Maize GMO Standard MON 863

for the detection of GMO Maize (MON 863) with PCR

DOWNLOAD MSDS (PDF)

Synonym: GMO Maize Powder Standard MON 863

Properties

Related Categories Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments, Food and Beverage,
CofA   certificate is enclosed in each package
quality   for the detection of GMO Maize (MON 863) with PCR
composition   genetically modified MON 863 maize, 1%
storage temp.   2-8°C

Price and Availability

Customers Also Viewed

for the detection of GMO Maize (MON 863) with PCR

for the detection of GMO Maize (MON 863) with PCR

for the detection of GMO Maize (MON 863) with PCR

Set for 0%, 0.1%, 1% and 9,9% MON 863

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?