Synonym: GMO Standard ERM-BF424
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, M, MA - MD More... |
| grade | BCR® certified Reference Material |
| CofA | certificate is enclosed in each package |
| composition | maize (59122), 10% |
BCR is a registered trademark of European Commission
Customers Also Viewed
10% 1507, for the detection of GMO Maize (1507) with PCR
BCR® certified Reference Material, for the detection of GMO Maize (59122) with PCR
for the detection of GMO Maize (MON 863) with PCR
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?