F2793 Fluka

D-(−)-Fructose

analytical standard for fructose assay kit, for use with enzymatic assay kit FA20

DOWNLOAD MSDS (PDF)

Synonym: D-Levulose, Fruit sugar

  • CAS Number 57-48-7

  • Empirical Formula (Hill Notation) C6H12O6

  • Molecular Weight 180.16

  •  Beilstein Registry Number 1239004

  •  EC Number 200-333-3

  •  MDL number MFCD00148910

  •  PubChem Substance ID 24894831

  • Popular Documents:  Bulletin (PDF)  

Price and Availability

Customers Also Viewed

<SC>D</SC>-(+) Glucose

analytical standard

<SC>D-</SC>(−)-Fructose

≥99%

<SC>D-</SC>(−)-Fructose

BioXtra

<SC>D-</SC>(−)-Fructose

meets USP testing specifications

sufficient for 20 assays

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?