H2016 Fluka

HPLC peptide standard mixture

DOWNLOAD MSDS (PDF)

Properties

Related Categories Alphabetical Index of Analytical Standards, Amino Acids, Peptides & Proteins, Analytical Standards, Analytical/Chromatography, Chromatography,
description   Peptide standard
composition   angiotensin II
  Gly-Tyr
  Leu enkephalin
  Met enkephalin
  Val-Tyr-Val
concentration   ~0.5 mg each component (dried film)
compatibility   use to QA Reversed phase columns
storage temp.   −20°C

Description

Packaging

Amber vials containing a dried film composed of approx. 0.5 mg each of Gly-Tyr, Val-Tyr-Val, methionine enkephalin, leucine enkephalin and angiotensin II.

Price and Availability

Customers Also Viewed

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?