-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
Synonym: α-Amylase from Bacillus amyloliquefaciens
| Related Categories | A, AM to AQ, Alphabetic, Analytical Standards, Analytical/Chromatography, |
| grade | BCR® certified Reference Material |
| shipped in | wet ice |
| storage temp. | −20°C |
BCR is a registered trademark of European Commission
Customers Also Viewed
aqueous solution, ≥800 FAU/g
powder, ~30 U/mg
Type XII-A, saline solution, 500-1,000 units/mg protein (biuret)
suitable for determination of starch (Kit STA-20)
lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?