Synonym:
N-(3-Oxohexanoyl)-
CAS Number 76924-95-3
Empirical Formula (Hill Notation) C10H15NO4
Molecular Weight 213.23
MDL number MFCD03788849
PubChem Substance ID 24896229
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Structure, Chromatography, |
| Quality Level | ELITE |
| grade | analytical standard |
| storage temp. | −20°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?