M2178 Fluka

Mycoplasma agar base

for microbiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories A - M, All Solid Media/Agars, Clinical Media, Media, Media by Application,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
final pH   7.8±0.2 (25 °C)
suitability   Mycoplasma

Description

Components

Ingredients (g/L)
Beef heart, infusion, 250.00
Peptic digest of animal tissue, 10.00
Sodium chloride, 5.00
Agar, 15.00

Application

Used for isolation and cultivation of mycoplasma.

Price and Availability

Customers Also Viewed

for microbiology

powder, suitable for microbiology

powder, suitable for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?