EMAIL THIS PAGE TO A FRIEND

M8802 Fluka

Malonate broth

for microbiology, pH 6.7±0.2 (25 °C)

Purchase

Description

Components

Components (g/L)
Ammonium sulfate, 2.00
Dipotassium phosphate, 0.60
Monopotassium phosphate, 0.40
Sodium chloride, 2.00
Sodium malonate, 3.00
Bromo thymol blue, 0.025

Application

Recommended for the differentiation of Enterobacter and Escherichia.

Price and Availability

Customers Also Viewed

for microbiology

for microbiology

for microbiology

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Ewing, W., et al. Publ. Hlth. Lab. 15, 153, (1957)

MacFaddin, J Media for Isolation-Cultivation-Identification-Maintenance of Medical Bacteria Baltimore, Maryland , (1985) 1

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?