EMAIL THIS PAGE TO A FRIEND

N4642 Fluka

N-Z-Case® Plus

Casein enzymatic hydrolysate, from bovine milk

Synonym: Peptone from casein

Purchase

Properties

Related Categories Media Components, Microbial Media for Molecular Biology and Protein Expression, Molecular Biology, Protein Hydrolysates
biological source   from bovine milk
nitrogen analysis   ≥6.0% amino, ≥10.0% total

Description

Other Notes

Enzymatic hydrolysis has been optimized to produce rich growth of most bacteria.

Packaging

1 kg in poly bottle

250 g in poly bottle

5 kg in poly drum

Reconstitution

Recommended concentration: 250 g/L

Application

Casein hydrolysates are used in the formulation of culture media for the cultivation of heterotrophic microorganisms. N-Z-Case® Plus is to be used in laboratory media and fermentation where a high degree of solution clarity is required.

Legal Information

N-Z-Case is a registered trademark of Kerry Group

Price and Availability

Customers Also Viewed

Casein acid hydrolysate, from bovine milk

Casein enzymatic hydrolysate, from bovine milk

for microbiology

Moderately digested casein enzymatic hydrolysate, from bovine milk

NZCYM Broth

EZMix powder microbial growth medium

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?