Synonym: Peptone from animal tissue
CAS Number 73049-73-7
MDL number MFCD00131829
Popular Documents: Specification Sheet (PDF)
| Related Categories | Core Bioreagents, Media Components, Microbial Media, Microbial Media for Molecular Biology, Molecular Biology, |
For the preparation of general culture media.
Bacteriological peptone contains 14.2 - 17.4% total nitrogen in a readily available form for bacteria. It also has a high peptide and amino acid content.
Customers Also Viewed
from meat, powder, suitable for cell culture
from meat, Type I, for microbiology
for microbiology
Enzymatic hydrolysate
Microbiologically tested.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?