CAS Number 113-92-8
Empirical Formula (Hill Notation) C16H19ClN2 · C4H4O4
Molecular Weight 390.86
EC Number 204-037-5
MDL number MFCD00069225
| Related Categories | Analytical Standards, Analytical/Chromatography, Chromatography, Pharmaceutical Secondary Standards, Pharmaceutical Standards More... |
| grade | certified reference material |
| packaging | pkg of 500 mg |
| mp | 130-135 °C(lit.) |
| pharmacopeia traceability | traceable to BP 81 |
| traceable to PhEur C1800000 | |
| traceable to USP 1123102 |
H1 Histamine receptor antagonist.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?