EMAIL THIS PAGE TO A FRIEND

PHR1051 Fluka

Propylene glycol

pharmaceutical secondary standard; traceable to USP

Synonym: 1,2-Propanediol, Propylene glycol

  • CAS Number 57-55-6

  • Linear Formula CH3CH(OH)CH2OH

  • Molecular Weight 76.09

  •  Beilstein Registry Number 1340498

  •  E Number E1520

  •  EC Number 200-338-0

  •  MDL number MFCD00064272

Purchase

Properties

Related Categories APIs & Metabolites, APIs (Active Pharmaceutical Ingredients) & Metabolites, Alcohol, All Alphabetically Sorted, Analytical Standards,
grade   certified reference material
vapor density   2.62 (vs air)
vapor pressure   0.08 mmHg ( 20 °C)
autoignition temp.   779 °F
expl. lim.   12.5 %
refractive index   n20/D 1.432(lit.)
bp   187 °C(lit.)
mp   −60 °C(lit.)
density   1.036 g/mL at 25 °C(lit.)
pharmacopeia traceability   traceable to USP 1576708

Description

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

General description

Certified Fluka® pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards

Other Notes

This product was designed, produced and verified for accuracy and stability in accordance with ISO/IEC 17025:2005 (AClass Cert AT-1467), ISO GUIDES 34:2009 (AClass Cert AR-1470).

Legal Information

FLUKA is a registered trademark of FLUKA GmbH

Price and Availability

Customers Also Viewed

1,2-Propanediol

puriss. p.a., ACS reagent, ≥99.5% (GC)

1,2-Propanediol

analytical standard

Propylene glycol

United States Pharmacopeia (USP) Reference Standard

Propylene glycol

≥99.5%, FCC, FG

Propylene glycol

European Pharmacopoeia (EP) Reference Standard

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
TY2000000
Flash Point(F) 
217.4 °F
Flash Point(C) 
103 °C

Protocols & Articles

Articles

Determination of Water Content in Propanediol Using Karl Fischer Titration

The determination of water in alcohols is straightforward. The substances dissolve very easily in methanol and addition of solubility mediators is only necessary in the case of long chain alcohols. I...
Keywords: Pharmaceutical, Titrations

Related Content

Flavor and Fragrance Analysis

It’s all about your workflow. Sample preparation, separation, detection and analysis. Speed and resolution is key in determining purity or verifying composition.
Keywords: Calorimetry, Gas chromatography, Mass spectrometry, Sample preparations, Separation

Reliable Identification of F&F Components by GC/MS and Linear Retention Indices and Multidimensional Chromatography [VIDEO]

In this on-demand webinar by Prof. Luigi Mondello of the University of Messina, Italy, highlights the shortcomings of GC/MS analysis in the identification of F&F compounds when using a straightforwar...
Keywords: Chromatography, Gas chromatography, High performance liquid chromatography, Mass spectrometry

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Engineering a cyanobacterium as the catalyst for the photosynthetic conversion of CO2 to 1,2-propanediol. Li H and Liao JC Microb. Cell Fact. 12, 4, (2013)

Loading...


HMDB: a knowledgebase for the human metabolome. Wishart DS, Knox C, Guo AC, et al. Nucleic Acids Res. 37(Database issue), D603-10, (2009)

Loading...


Recognition, treatment, and prevention of propylene glycol toxicity. Zar T, Graeber C, and Perazella MA Semin. Dial. 20(3), 217-9, (2007)

Loading...


Terpenes in propylene glycol as skin-penetration enhancers: permeation and partition of haloperidol, Fourier transform infrared spectroscopy, and differential scanning calorimetry. Vaddi HK, Ho PC, and Chan SY J. Pharm. Sci. 91(7), 1639-51, (2002)

Loading...


Understanding the human salivary metabolome. Takeda I, Stretch C, Barnaby P, et al. NMR Biomed. 22(6), 577-84, (2009)

Loading...


The human cerebrospinal fluid metabolome. Wishart DS, Lewis MJ, Morrissey JA, et al. J. Chromatogr. B. Analyt. Technol. Biomed. Life Sci. 871(2), 164-73, (2008)

Loading...


Prediction of skeletal muscle and fat mass in patients with advanced cancer using a metabolomic approach. Stretch C, Eastman T, Mandal R, et al. J. Nutr. 142(1), 14-21, (2012)

Loading...


From kinetic-structure analysis to engineering crystalline fiber networks in soft materials. Wang RY, Wang P, Li JL, et al. Phys. Chem. Chem. Phys. 15(9), 3313-9, (2013)

Loading...


In vitro iontophoresis of R-apomorphine across human stratum corneum. Structure-transport relationship of penetration enhancement. Li GL, van der Geest R, Chanet L, et al. J. Control. Release 84(1-2), 49-57, (2002)

Loading...


The influence of self-assembling supramolecular structures on the passive membrane transport of ion-paired molecules. Benaouda F, Brown MB, Shah B, et al. Int. J. Pharm. 439(1-2), 334-41, (2012)

Loading...


1H and (13)C NMR spectroscopic analysis of human saliva. Silwood CJ, Lynch E, Claxson AW, et al. J. Dent. Res. 81(6), 422-7, (2002)

Loading...


Role of complement inhibition in topical therapy of muco-cutaneous herpes simplex virus infections. Gancevici GG Roum. Arch. Microbiol. Immunol. 52(4), 293-303, (1993)

Loading...


Systemic contact dermatitis due to intravenous Valium in a person sensitive to propylene glycol. Fisher AA Cutis. 55(6), 327-8, (1995)

Loading...


[Intramuscular premedication with diazepam in a fat emulsion]. Faré JC, Guesnon P, Helouis JJ, et al. Cah. Anesthesiol. 32(4), 303-6, (1984)

Loading...


[Analysis of methionine metabolism studied by the gas chromatographic determination of 3-methylthiopropionate in urine and its clinical application]. Yoshida Y Hokkaido Igaku Zasshi. 60(2), 183-94, (1985)

Loading...


The measurement of 1,2-propanediol, D, L-2,3-butanediol and meso-2,3-butanediol in controls and alcoholic cirrhotics. Casazza JP, Frietas J, Stambuk D, et al. Alcohol Alcohol Suppl. 1, 607-9, (1987)

Loading...


Simultaneous determination of plasma phenethylamine, phenylethanolamine, tyramine and octopamine by high-performance liquid chromatography using derivatization with fluorescamine. Yonekura T, Kamata S, Wasa M, et al. J. Chromatogr. A 427(2), 320-5, (1988)

Loading...


Hidden formaldehyde in e-cigarette aerosols. Jensen RP, Luo W, Pankow JF, et al. N. Engl. J. Med. 372(4), 392-4, (2015)

Loading...


Nonacetogenic growth of the acetogen Acetobacterium woodii on 1,2-propanediol. Schuchmann K, Schmidt S, Martinez Lopez A, et al. J. Bacteriol. 197(2), 382-91, (2015)

Loading...


E-cigarettes: The lingering questions. Cressey D Nature 513(7516), 24-6, (2014)

Loading...


Are e-cigarettes safe? Maron DF Sci. Am. 310(5), 31-2, (2014)

Loading...


Positive propylene glycol result in a patient with ethylene glycol poisoning. Su Z, Stone RW, and Zhu Y Clin. Chem. 60(4), 697-8, (2014)

Loading...


Quiz page October 2013: When things are not as they seem: a woman with anion gap metabolic acidosis after ingesting antifreeze. Alavala A, Ulliman E, Shirazi M, et al. Am. J. Kidney Dis. 62(4), A25-8, (2013)

Loading...


Fit of fluxes of sunscreens and other compounds from propylene glycol:water (30:70) through human skin and silicone membrane to the Roberts-Sloan equation: the effect of polar vehicle (or water) solubility. Sloan KB, Devarajan-Ketha H, Synovec J, et al. J. Cosmet. Sci. 64(3), 181-92, (2013)

Loading...


Metabolomic analysis of cerebral spinal fluid from patients with severe brain injury. Glenn TC, Hirt D, Mendez G, et al. Acta Neurochir. Suppl. 118, 115-9, (2013)

Loading...


Effect of intratympanic mitomycin C on the development of cholesteatoma and otitis media in rats. Melo AA, Caldas Neto SS, Leão FS, et al. J. Laryngol. Otol. 127(4), 359-63, (2013)

Loading...


Inhibition of skin inflammation in mice by diclofenac in vesicular carriers: liposomes, ethosomes and PEVs. Caddeo C, Sales OD, Valenti D, et al. Int. J. Pharm. 443(1-2), 128-36, (2013)

Loading...


Enhancement of anti-cholinesterase activity of Zingiber cassumunar essential oil using a microemulsion technique. Okonogi S and Chaiyana W Drug Discov. Ther. 6(5), 249-55, (2012)

Loading...


Antibacterial action of calcium hydroxide vehicles and calcium hydroxide pastes. Pacios MG, Silva C, López ME, et al. J. Investig. Clin. Dent. 3(4), 264-70, (2012)

Loading...


Safety assessment of propylene glycol, tripropylene glycol, and PPGs as used in cosmetics. Fiume MM, Bergfeld WF, Belsito DV, et al. Int. J. Toxicol. 31(5 Suppl), 245S-60S, (2012)

Loading...


Skin-sensitizing and irritant properties of propylene glycol. Lessmann H, Schnuch A, Geier J, et al. Contact Dermatitis 53(5), 247-59, (2005)

Loading...


[Study on the toxicological effect of chloropropanols on rats]. Li N, Liu Z, Jia X, et al. Wei Sheng Yan Jiu 32(4), 349-52, (2003)

Loading...


The phase diagram for the ternary system propylene glycol-sodium chloride-water and their application to platelet cryopreservation. Liu JH, Gao D, He LQ, et al. Zhongguo Shi Yan Xue Ye Xue Za Zhi 11(1), 92-5, (2003)

Loading...


Microbial formation, biotechnological production and applications of 1,2-propanediol. Bennett GN and San KY Appl. Microbiol. Biotechnol. 55(1), 1-9, (2001)

Loading...


The chicken type III GnRH receptor homologue is predominantly expressed in the pituitary, and exhibits similar ligand specificity to the type I receptor. Joseph, N.T., et al. J. Endocrinol. 202, 179, (2009)

Loading...


The human serum metabolome. Psychogios N, Hau DD, Peng J, et al. PLoS ONE 6(2), e16957, (2011)

Loading...


Epirubicin loaded with propylene glycol liposomes significantly overcomes multidrug resistance in breast cancer. Zhao YZ, Dai DD, Lu CT, et al. Cancer Lett. 330(1), 74-83, (2013)

Loading...


GnRH receptor activation competes at a low level with growth signaling in stably transfected human breast cell lines. Morgan, K BMC Cancer 11, 476, (2011)

Loading...


The use of an open-freezing system with self-seeding for cryopreservation of mouse ovarian tissue. Maltaris T, Dimmler A, Müller A, et al. Reprod. Domest. Anim. 40(3), 250-4, (2005)

Loading...


Hydrogen-driven asymmetric reduction of hydroxyacetone to (R)-1,2-propanediol by Ralstonia eutropha transformant expressing alcohol dehydrogenase from Kluyveromyces lactis. Oda T, Oda K, Yamamoto H, et al. Microb. Cell Fact. 12(1), 2, (2013)

Loading...


The PduQ enzyme is an alcohol dehydrogenase used to recycle NAD+ internally within the Pdu microcompartment of Salmonella enterica. Cheng S, Fan C, Sinha S, et al. PLoS ONE 7(10), e47144, (2012)

Loading...


Cryopreservation of rabbit semen: comparing the effects of different cryoprotectants, cryoprotectant-free vitrification, and the use of albumin plus osmoprotectants on sperm survival and fertility after standard vapor freezing and vitrification. Rosato MP and Iaffaldano N Theriogenology 79(3), 508-16, (2013)

Loading...


High-resolution proton magnetic resonance analysis of human cerebrospinal fluid. Petroff OA, Yu RK, and Ogino T J. Neurochem. 47(4), 1270-6, (1986)

Loading...


Serum concentration of 7 alpha-hydroxycholesterol as an indicator of bile acid synthesis in humans. Hahn C, Reichel C, and von Bergmann K J. Lipid Res. 36(9), 2059-66, (1995)

Loading...


Propylene glycol-induced proximal renal tubular cell injury. Yorgin PD, Theodorou AA, Al-Uzri A, et al. Am. J. Kidney Dis. 30(1), 134-9, (1997)

Loading...


Oxide terpenes as human skin penetration enhancers of haloperidol from ethanol and propylene glycol and their modes of action on stratum corneum. Vaddi HK, Ho PC, Chan YW, et al. Biol. Pharm. Bull. 26(2), 220-8, (2003)

Loading...


Characterization of serum and urinary bile acids in patients with primary biliary cirrhosis by gas-liquid chromatography-mass spectrometry: effect of ursodeoxycholic acid treatment. Batta AK, Arora R, Salen G, et al. J. Lipid Res. 30(12), 1953-62, (1989)

Loading...


A review of the comparative mammalian toxicity of ethylene glycol and propylene glycol. LaKind JS, McKenna EA, Hubner RP, et al. Crit. Rev. Toxicol. 29(4), 331-65, (1999)

Loading...


Plasma amino acid levels with a note on membrane transport: characteristics, regulation, and metabolic significance. Cynober LA Nutrition 18(9), 761-6, (2002)

Loading...


Influence of penetration enhancer on drug permeation from volatile formulations. Santos P, Watkinson AC, Hadgraft J, et al. Int. J. Pharm. 439(1-2), 260-8, (2012)

Loading...


A new paradigm in dermatopharmacokinetics - Confocal Raman spectroscopy. Mateus R, Abdalghafor H, Oliveira G, et al. Int. J. Pharm. 444(1-2), 106-8, (2013)

Loading...


Novel double phase transforming organogel based on β-cyclodextrin in 1,2-propylene glycol. Liu W, Xing P, Xin F, et al. J. Phys. Chem. B 116(43), 13106-13, (2012)

Loading...


Blood optical clearing studied by optical coherence tomography. Zhernovaya O, Tuchin VV, and Leahy MJ J. Biomed. Opt. 18(2), 26014, (2013)

Loading...


LC analysis of benzophenone-3: II application to determination of 'in vitro' and 'in vivo' skin penetration from solvents, coarse and submicron emulsions. Fernandez C, Marti-Mestres G, Ramos J, et al. J. Pharm. Biomed. Anal. 24(1), 155-65, (2000)

Loading...


Impact of flavour solvent (propylene glycol or triacetin) on vanillin, 5-(hydroxymethyl)furfural, 2,4-decadienal, 2,4-heptadienal, structural parameters and sensory perception of shortcake biscuits over accelerated shelf life testing. Yang N, Hort J, Linforth R, et al. Food Chem. 141(2), 1354-60, (2013)

Loading...


Real time and non-destructive analysis of tablet coating thickness using acoustic microscopy and infrared diffuse reflectance spectroscopy. Bikiaris D, Koutri I, Alexiadis D, et al. Int. J. Pharm. 438(1-2), 33-44, (2012)

Loading...


The effects of cryosolvents on DOPC-β-sitosterol bilayers determined from molecular dynamics simulations. Hughes ZE, Malajczuk CJ, and Mancera RL J. Phys. Chem. B 117(12), 3362-75, (2013)

Loading...


The effect of ultrasound on the in vitro penetration of ibuprofen through human epidermis. Brucks R, Nanavaty M, Jung D, et al. Pharm. Res. 6(8), 697-701, (1989)

Loading...


Propylene glycol and glycerol concentration in ultrasound gel. Belavy D and Wallis SC Reg. Anesth. Pain Med. 38(1), 75-6, (2013)

Loading...


Glucogenic supply increases oocyte developmental competence in sheep. Berlinguer F, Gonzalez-Bulnes A, Contreras-Solis I, et al. Reprod. Fertil. Dev. 24(8), 1055-62, (2012)

Loading...


A monoclonal antibody-based enzyme-linked immunosorbent assay of glycolithocholic acid sulfate in human urine for liver function test. Kobayashi N, Katsumata H, Uto Y, et al. Steroids 67(10), 827-33, (2002)

Loading...


Propylene glycol dermatitis: re-evaluation of an old problem. Funk JO and Maibach HI Contact Dermatitis 31(4), 236-41, (1994)

Loading...


Expression and inducibility of antigens in severe combined immunodeficient mice recognized by human anti-P450 antibodies. Rajasenan RS, Riley RJ, and Leeder JS Toxicol. Appl. Pharmacol. 135(1), 89-99, (1995)

Loading...


Are all aciclovir cream formulations bioequivalent? Trottet L, Owen H, Holme P, et al. Int. J. Pharm. 304(1-2), 63-71, (2005)

Loading...


Modeling the impact of solid surfaces in thermal degradation processes. Tuma C, Laino T, Martin E, et al. ChemPhysChem 14(1), 88-91, (2013)

Loading...


Exposure of German residents to ethylene and propylene glycol ethers in general and after cleaning scenarios. Fromme H, Nitschke L, Boehmer S, et al. Chemosphere 90(11), 2714-21, (2013)

Loading...


Blood methanethiol in alcoholic liver disease with and without hepatic encephalopathy. McClain CJ, Zieve L, Doizaki WM, et al. Gut 21(4), 318-23, (1980)

Loading...


Liquisolid technique: a promising alternative to conventional coating for improvement of drug photostability in solid dosage forms. Khames A Expert Opin. Drug Deliv. 10(10), 1335-43, (2013)

Loading...


In vivo and in vitro evaluation of the estrogenic properties of the 17β-(butylamino)-1,3,5(10)-estratrien-3-ol (buame) related to 17β-estradiol. Lemini C, Medina M, Avila ME, et al. Pharmacol. Rep. 64(4), 940-50, (2012)

Loading...


Guanidino compounds in serum and urine of cirrhotic patients. Marescau B, De Deyn PP, Holvoet J, et al. Metabolism 44(5), 584-8, (1995)

Loading...


Selection of high efficient transdermal lipid vesicle for curcumin skin delivery. Zhao YZ, Lu CT, Zhang Y, et al. Int. J. Pharm. 454(1), 302-9, (2013)

Loading...


Molecular dynamics simulations of the interactions of DMSO, mono- and polyhydroxylated cryosolvents with a hydrated phospholipid bilayer. Malajczuk CJ, Hughes ZE, and Mancera RL Biochim. Biophys. Acta 1828(9), 2041-55, (2013)

Loading...


Acetone metabolism in humans during diabetic ketoacidosis. Reichard GA, Skutches CL, Hoeldtke RD, et al. Diabetes 35(6), 668-74, (1986)

Loading...


Higher levels of plasma cholesterol sulfate in patients with liver cirrhosis and hypercholesterolemia. Tamasawa N, Tamasawa A, and Takebe K Lipids 28(9), 833-6, (1993)

Loading...


Percutaneous absorption and distribution of organophosphates (chlorpyrifos and dichlorvos) following dermal exposure and decontamination scenarios using in vitro human skin model. Moore CA, Wilkinson SC, Blain PG, et al. Toxicol. Lett. 229(1), 66-72, (2014)

Loading...


Use of a human skin in vitro model to investigate the influence of 'every-day' clothing and skin surface decontamination on the percutaneous penetration of organophosphates. Moore CA, Wilkinson SC, Blain PG, et al. Toxicol. Lett. 229(1), 257-64, (2014)

Loading...


Plasma leucine enkephalin is increased in liver disease. Thornton JR and Losowsky MS Gut 30(10), 1392-5, (1989)

Loading...


A new method for determination of serum cholestanol by high-performance liquid chromatography with ultraviolet detection. Halperin G, Elisaf M, Leitersdorf E, et al. J. Chromatogr. B. Biomed. Sci. Appl. 742(2), 345-52, (2000)

Loading...


Thermoanalytical probes for the analysis of physical properties of stratum corneum. Miller DL and Wildnauer RH J. Invest. Dermatol. 69(3), 287-9, (1977)

Loading...


Plasma fatty acid pattern including diene-conjugated linoleic acid in ethanol users and patients with ethanol-related liver disease. Szebeni J, Eskelson C, Sampliner R, et al. Alcohol. Clin. Exp. Res. 10(6), 647-50, (1986)

Loading...


Evaluation of activated sludge for biodegradation of propylene glycol as an aircraft deicing fluid. Delorit JD and Racz L Water Environ. Res. 86(4), 366-71, (2014)

Loading...


Influence of nanoparticle concentration on thermo-physical properties of CuO-propylene glycol nanofluids. Suganthi KS, Radhakrishnan AK, Anusha N, et al. J. Nanosci. Nanotechnol. 14(6), 4602-7, (2014)

Loading...


Transcutaneous immunization system using a hydrotropic formulation induces a potent antigen-specific antibody response. Takatani-Nakase T, Tokuyama E, Komai M, et al. PLoS ONE 7(10), e47980, (2012)

Loading...


Merck 14,7855

Beil. 1,IV,2468

Aldrich MSDS 1, 1539:C / Arctander, 2696 / Corp MSDS 1 (2), 2949:A / FT-IR 2 (1), 186:D / FT-IR 1 (1), 128:B / FT-NMR 1 (1), 192:C / IR-Spectra (3), 80:B / IR-Spectra (2), 77:F / NMR-Reference 2 (1), 120:B / RegBook 1 (1), 139:C / RegBook 96 (1), 139:C / Sax 6, 2293 / Sigma FT-IR 1 (2), 862:A / Structure Index 1, 16:D:6 / Vapor Phase 3, 188:D

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?