PHR1118 Fluka

Propyl gallate

pharmaceutical secondary standard; traceable to USP and PhEur

DOWNLOAD MSDS (PDF)

Synonym: 3,4,5-Trihydroxybenzoic acid propyl ester, Tenox PG

  • CAS Number 121-79-9

  • Linear Formula 3,4,5-(HO)3C6H2CO2CH2CH2CH3

  • Molecular Weight 212.20

  •  Beilstein Registry Number 1877976

  •  E Number E310

  •  EC Number 204-498-2

  •  MDL number MFCD00002196

Properties

Related Categories All Alphabetically Sorted, Analytical Standards, Analytical/Chromatography, Chemical Class, Chromatography,
grade   certified reference material
packaging   pkg of 1 g
mp   146-149 °C(lit.)
pharmacopeia traceability   traceable to PhEur P3640000
  traceable to USP 1576800

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO Guide 34.

Other Notes

Values of analytes vary lot to lot.

Reconstitution

Prepare a 0.1M solution in glycerol:PBS (9:1)

Application

Used as an anti-fade reagent in fluorescence microscopy to reduce photobleaching of fluorescent probes such as rhodamine and fluorescein.

Biochem/physiol Actions

An antioxidant that exhibits antimicrobial activity. Propyl gallate has been reported to be an effective antioxidant-based hepatoprotector, both in vitro and in vivo. It has also been shown to prevent neuronal apoptosis and block the death of neurons exposed to FeSO4/GA as well as partially protect endothelial cells against TNF-induced apoptosis. However, propyl gallate induced single strand breaks in DNA at concentrations higher than 0.25 μM when it was combined with copper concentrations at 5 μM and above. Another report, however, found that incubation of isolated rat hepatocytes with propyl gallate at concentrations of >1 mM induced cell killing, whereas, a lower concentration of propyl gallate > 0.5 mM resulted in a ladder formation of soluble low-molecular weight DNA fragments, characteristic of apoptosis.

An antioxidant that exhibits antimicrobial activity. Propyl gallate has been reported to be an effective antioxidant-based hepatoprotector, both in vitro and in vivo. It has also been shown to prevent neuronal apoptosis and block the death of neurons exposed to FeSO4/GA as well as partially protect endothelial cells against TNF-induced apoptosis.

Price and Availability

Customers Also Viewed

Propyl gallate

powder

Propyl gallate

purum, antioxidant, ≥98.0% (HPLC)

Propyl gallate

FCC, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?