Synonym:
4-Amino-N-
CAS Number 723-46-6
Empirical Formula (Hill Notation) C10H11N3O3S
Molecular Weight 253.28
Beilstein Registry Number 6732984
EC Number 211-963-3
MDL number MFCD00010546
| Related Categories | APIs (Active Pharmaceutical Ingredients) & Metabolites, All alphabetically sorted, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | certified reference material |
| packaging | pkg of 1 g |
| pharmacopeia traceability | traceable to BP 314 |
| traceable to PhEur S2100000 | |
| traceable to USP 1631001 | |
| storage temp. | room temp |
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO Guide 34.
Values of analytes vary lot to lot.
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?