Beilstein Registry Number 1915399
MDL number MFCD00003654
PubChem Substance ID 24900279
Popular Documents: Datasheet (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Bioactive Small Molecules, Cell Biology, |
| grade | drug standard |
| drug control | Home Office Schedule 4.2 |
| concentration | 1.0 mg/mL±2% in 1,2-dimethoxyethane |
| storage temp. | 2-8°C |
| Gene Information |
human ... AR(367), EBP(10682), ESR1(2099), PGR(5241), SERPINA6(866), SHBG(6462) mouse ... Esr1(13982) rat ... Ar(24208), Esr1(24890), Nr3c1(24413), Nr3c2(25672), Pgr(25154) |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?