Synonym:
5-
CAS Number 53885-35-1
Empirical Formula (Hill Notation) C14H14ClNS · HCl
Molecular Weight 300.25
EC Number 258-837-4
MDL number MFCD00079606
PubChem Substance ID 24900396
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, Drugs & Metabolites, |
| grade | analytical standard, for drug analysis |
| assay | ≥99% |
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?