EMAIL THIS PAGE TO A FRIEND

T7293 Fluka

Tryptone

Microbiologically tested.

Synonym: Peptone from casein

Purchase

Description

Preparation Note

Pancreatic digest of casein.

Application

Tryptone-yeast extract-glucose medium may be used in response surface methodology in order to find optimimum conditions for the growth of Lactobacillus casei YIT 9018.1 It was also used in the media used in the growth experiments with Escherichia coli MG165, conducted in order to monitor the bacteria for long term which underwent programmed population control, in a microchemostat, in an approach to prevent biofilm formation.2 Tryptone glucose extract agar (TGEA) along with gypsum board may also be used to cultivate Streptomyces albidoflavui in order to produce volatile metabolites. Tryptone may also be used in STGG (skim milk, tryptone, glucose, glycerol) medium at -80°C, which is used to preserve Streptococcus pneumoniae, Haemophilus influenzae, and Moraxella catarrhalis isolates for atleast 3 years.3

Price and Availability


Oxford Genetics

Customers Also Viewed

Tryptone

Pancreatic digest of casein

for microbiology

for biotechnological purposes

for microbiology

for use in microbial growth medium

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?