Synonym: Peptone from casein
CAS Number 91079-40-2
EC Number 293-428-4
| Related Categories | Broth / Media, Core Bioreagents, Life Science Reagents for Cloning, Media Components, Microbial Media, |
| nitrogen analysis | 11.4-13.9% total |
Used to supplement media in the cultivation of anaerobes (Clostridia and other fermenting organisms, e.g. Lactobacilli).
Customers Also Viewed
Pancreatic digest of casein
Microbiologically tested.
for microbiology
for biotechnological purposes
for microbiology
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?