• USA Home
  • Y0000156 - Tramadol impurity A

EMAIL THIS PAGE TO A FRIEND

Y0000156 Fluka

Tramadol impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym: (1RS,2SR)-2-[(Dimethylamino)methyl]-1-(3-methoxyphenyl)cyclohexanol

  • Empirical Formula (Hill Notation) C16H25NO2

  • Molecular Weight 263.38

Purchase

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Other Notes

Sales restrictions may apply.

Packaging

Unit quantity: 25 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference subtance catalogue.

Price and Availability


Explore the New Analytical Standards Portal

Try now and save 50%

Customers Also Viewed

Paracetamol
P0300000Paracetamol

European Pharmacopoeia (EP) Reference Standard

Tramadol hydrochloride

≥99.0% (HPLC)

Tramadol hydrochloride

≥98% (HPLC), solid

European Pharmacopoeia (EP) Reference Standard

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?