• USA Home
  • 1.03834 - Fittings for capillaries with 3.2 mm O.D., for HPLC bottle adapter S40

EMAIL THIS PAGE TO A FRIEND
1.03834 EMD Millipore

Fittings for capillaries with 3.2 mm O.D., for HPLC bottle adapter S40

(pack of 10)

Purchase

Properties

packaging   10 st in Plastic bag (1038340001)
shipped in   ambient

Description

Other Notes

These fittings are spare parts for our HPLC bottle-adapters 1.03830.0001 and 1.03831.0001 to fix the HPLC-tubes in the connection openings by thread. Material: PP blue

Price and Availability

Safety & Documentation

Safety Information

HS Code 
3926 90 97
Storage class 
10 - 13 Other liquids and solids

Documents

Certificate of Analysis

Protocols & Articles

Articles

Safety Accessories

Safety Accessories for Bottles Bottles made from glass, aluminum, and HDPE are widely used for packaging and storing solvents and inorganics. We provide a broad range of specially developed safety ac...
Keywords: Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?