• USA Home
  • 4.90293 - 1,3-Dimethylimidazolium dimethylphosphate

EMAIL THIS PAGE TO A FRIEND
4.90293 EMD Millipore

1,3-Dimethylimidazolium dimethylphosphate

1,3-Dimethylimidazolium dimethylphosphate for synthesis. CAS 654058-04-5, pH 2.4 (H₂O, 20 °C)., for synthesis

  • CAS Number 654058-04-5

  • Linear Formula C₇H₁₅N₂O₄P

  • Molecular Weight 222.18 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (4902930025)
density   1.27 g/cm3 (20 °C)
pH   2.4 (H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Purity (HPLC): ≥ 98.0 %
  Identity (NMR): passes test
  Halides (IC): ≤ 0.1 %
  Water (K. F.): ≤ 1.0 %

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 29 90
Hazard Statement(s) 
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 1760 , 8, II
Declaration (transport by air) IATA-DGR 
UN 1760 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 1760 , 8, II
Categories of danger 
corrosive
R Phrase 
R 34
Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?